siRNA Details
Gene Symbol: Six3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015385.2 | 2 | 533 | |||
NR_015386.2 | 2 | 442 | ||||
NR_015387.2 | 3 | 795 | ||||
NR_038082.1 | 2 | 442 | ||||
NR_038083.1 | 2 | 442 | ||||
NR_038084.1 | 2 | 442 | ||||
NR_038085.1 | 2 | 442 | ||||
NR_038086.1 | 2 | 442 | ||||
GenBank mRNA | AK044330.1 | - | LIT3371 RNCR1 |
2 | 2304 | |
AK053722.1 | - | 2 | 444 | |||
AY589790.1 | - | 2 | 321 | |||
AY589791.1 | - | 3 | 764 | |||
AY589792.1 | - | 1457 | ||||
AY590888.1 | - | 2 | 271 | |||
AY590889.1 | - | 2 | 361 | |||
BC065087.1 | - | 2 | 431 | |||
CA527570.1 | - | 2 | 386 | |||
CF737515.1 | - | 2 | 402 | |||
CF737925.1 | - | 2 | 418 |
