siRNA Details
Gene Symbol: Six3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015385.2 | 1 | 134 | |||
NR_015386.2 | 1 | 134 | ||||
NR_015387.2 | 1 | 134 | ||||
NR_038082.1 | 1 | 134 | ||||
NR_038083.1 | 1 | 134 | ||||
NR_038084.1 | 1 | 134 | ||||
NR_038085.1 | 1 | 134 | ||||
NR_038086.1 | 1 | 134 | ||||
GenBank mRNA | AK044330.1 | - | LIT3371 RNCR1 |
1 | 104 | |
AK053722.1 | - | 1 | 136 | |||
AY589790.1 | - | 1 | 13 | |||
AY589791.1 | - | 1 | 103 | |||
AY589792.1 | - | 1149 | ||||
BC065087.1 | - | 1 | 123 | |||
CA527570.1 | - | 1 | 78 | |||
CF737515.1 | - | 1 | 94 | |||
CF737925.1 | - | 1 | 110 |
