siRNA Details
Gene Symbol: Gm11978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028586.1 | 2 | 169 | |||
GenBank mRNA | AK132798.1 | - | 2 | 170 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614890.1 | - | 2 | 166 | Chr. 11 - 6650844 - 6660233 [-] on Build GRCm38 |
