siRNA Details
Gene Symbol: Gm4532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030674.1 | 1 | 696 | |||
GenBank mRNA | AK011173.1 | - | 1 | 698 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX524884.1 | - | 1 | 455 | Chr. 7 - 127232423 - 127232889 [-] on Build GRCm38 |
