siRNA Details
Gene Symbol: Gm4532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030674.1 | 1 | 652 | |||
GenBank mRNA | AK011173.1 | - | 1 | 654 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030674.1 | 1 | 652 | |||
GenBank mRNA | AK011173.1 | - | 1 | 654 |