siRNA Details
Gene Symbol: Pinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003202.1 | 1 | 36 | |||
GenBank mRNA | DQ059755.1 | - | LIT3460 PINC |
1 | 36 | |
DQ059756.1 | - | LIT3461 PINC |
1 | 36 |
