siRNA Details
Gene Symbol: Pinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003202.1 | 5 | 1439 | |||
GenBank mRNA | DQ059755.1 | - | LIT3460 PINC |
5 | 1439 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003202.1 | 5 | 1439 | |||
GenBank mRNA | DQ059755.1 | - | LIT3460 PINC |
5 | 1439 |