siRNA Details
Gene Symbol: 4930417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015527.1 | 3, 4 | 762 | |||
GenBank mRNA | AK037571.1 | - | 3, 4 | 669 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB155665.2 | - | 3, 4 | 346 | Chr. 6 - 125265946 - 125273777 [+] on Build GRCm38 |
