siRNA Details
Gene Symbol: 4930417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015527.1 | 1 | 435 | |||
GenBank mRNA | AK015159.1 | - | 1 | 436 | ||
AK029635.1 | - | 1 | 436 | |||
AK037571.1 | - | 1 | 342 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015527.1 | 1 | 435 | |||
GenBank mRNA | AK015159.1 | - | 1 | 436 | ||
AK029635.1 | - | 1 | 436 | |||
AK037571.1 | - | 1 | 342 |