siRNA Details
Gene Symbol: 4930417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015527.1 | 1 | 481 | |||
GenBank mRNA | AK015159.1 | - | 1 | 482 | ||
AK029635.1 | - | 1 | 482 | |||
AK037571.1 | - | 1 | 388 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB630781.1 | - | 1 | 385 | Chr. 6 - 125265621 - 125270398 [+] on Build GRCm38 |
