siRNA Details
Gene Symbol: 4930417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015527.1 | 3 | 733 | |||
GenBank mRNA | AK030803.1 | - | 1762 | |||
AK037571.1 | - | 3 | 640 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015527.1 | 3 | 733 | |||
GenBank mRNA | AK030803.1 | - | 1762 | |||
AK037571.1 | - | 3 | 640 |