siRNA Details
Gene Symbol: Pvt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003368.1 | 9 | 2577 | |||
GenBank mRNA | AK010708.1 | - | 1 | 622 | ||
Z12002.1 | - | 4 | 1671 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK169927.1 | - | 2300 | Chr. - - [-] on Build |
