siRNA Details
Gene Symbol: C030016D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027987.1 | 1 | 806 | |||
GenBank mRNA | AK047713.1 | - | 1 | 772 | ||
BC094907.1 | - | 1 | 806 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C030016D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027987.1 | 1 | 806 | |||
GenBank mRNA | AK047713.1 | - | 1 | 772 | ||
BC094907.1 | - | 1 | 806 |