siRNA Details
Gene Symbol: E330013P04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026942.1 | 5 | 1180 | |||
GenBank mRNA | AK087735.1 | - | 5 | 1166 | ||
BC076607.1 | - | 5 | 1180 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E330013P04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026942.1 | 5 | 1180 | |||
GenBank mRNA | AK087735.1 | - | 5 | 1166 | ||
BC076607.1 | - | 5 | 1180 |