siRNA Details
Gene Symbol: A430093F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027805.1 | 2 | 405 | |||
GenBank mRNA | AK079853.1 | - | 2 | 406 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_207529.1 | - | 406 | Chr. - - [-] on Build |
