siRNA Details
Gene Symbol: Gm5485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015373.1 | 4 | 589 | |||
GenBank mRNA | AK078896.1 | - | 4 | 590 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV374024.2 | - | 3 | 642 | Chr. 16 - 48399453 - 48412287 [-] on Build GRCm38 |
