siRNA Details
Gene Symbol: Gm5485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015373.1 | 2, 3 | 78 | |||
GenBank mRNA | AK078896.1 | - | 2, 3 | 79 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB582828.2 | - | 2, 3 | 75 | Chr. 16 - 48399450 - 48412413 [-] on Build GRCm38 |
