siRNA Details
Gene Symbol: 4930426L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024323.1 | 1 | 433 | |||
GenBank mRNA | AK015207.1 | - | 1 | 410 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930426L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024323.1 | 1 | 433 | |||
GenBank mRNA | AK015207.1 | - | 1 | 410 |