siRNA Details
Gene Symbol: 4930426L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024323.1 | 1 | 1391 | |||
GenBank mRNA | AK015207.1 | - | 1 | 1368 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB097091.1 | - | 1 | 222 | Chr. 2 - 18999492 - 18999802 [+] on Build GRCm38 |
