siRNA Details
Gene Symbol: A730017L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015523.2 | 2 | 307 | |||
GenBank mRNA | AK158456.1 | - | 2 | 308 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ142577.1 | - | 2 | 308 | Chr. 2 - 130874293 - 130906396 [-] on Build GRCm38 |
