siRNA Details
Gene Symbol: Hoxaas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015611.2 | 3 | 584 | |||
GenBank mRNA | AK012572.1 | - | 3 | 517 | ||
AK028207.1 | - | 3 | 585 | |||
AK146012.1 | - | 479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY737288.1 | - | 3 | 479 | Chr. 6 - 52175233 - 52213529 [+] on Build GRCm38 |

More Information
Other Species Targeted

Gene Ontology Categories:
