siRNA Details
Gene Symbol: Nctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002452.2 | 2 | 341 | |||
GenBank mRNA | AK035596.1 | - | 2 | 340 | ||
AK036720.1 | - | 2 | 341 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA062189.1 | - | 2 | 342 | Chr. 7 - 142555870 - 142558594 [-] on Build GRCm38 |
