siRNA Details
Gene Symbol: Smim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007443.1 | - | 1 | 149 | ||
AK019298.1 | - | 1 | 124 | |||
AK036279.1 | - | 112 | ||||
CX238554.1 | - | 131 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007443.1 | - | 1 | 149 | ||
AK019298.1 | - | 1 | 124 | |||
AK036279.1 | - | 112 | ||||
CX238554.1 | - | 131 |