siRNA Details
Gene Symbol: Mir7-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029825.1 | 1 | 49 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA881786.1 | - | 1 | 100 | Chr. 13 - 58392382 - 58392937 [-] on Build GRCm38 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NR_031795.
Gene Ontology Categories:
