siRNA Details
Gene Symbol: B330016D10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030695.1 | 1 | 1997 | |||
GenBank mRNA | AK046532.1 | - | 1 | 1993 | ||
AK148126.1 | - | 1 | 1345 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B330016D10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030695.1 | 1 | 1997 | |||
GenBank mRNA | AK046532.1 | - | 1 | 1993 | ||
AK148126.1 | - | 1 | 1345 |