siRNA Details
Gene Symbol: B330016D10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030695.1 | 1 | 59 | |||
GenBank mRNA | AK046111.1 | - | 1 | 59 | ||
AK046532.1 | - | 1 | 55 | |||
AK080800.1 | - | 1 | 63 | |||
AK142965.1 | - | 1 | 53 | |||
BB644677.1 | - | 1 | 59 |
