siRNA Details
Gene Symbol: B330016D10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030695.1 | 1 | 872 | |||
GenBank mRNA | AK043314.1 | - | 1 | 243 | ||
AK046111.1 | - | 1 | 872 | |||
AK046532.1 | - | 1 | 868 | |||
AK080800.1 | - | 1 | 879 | |||
AK080864.1 | - | 1 | 848 | |||
AK082239.1 | - | 1 | 250 | |||
AK142965.1 | - | 1 | 866 | |||
AK148126.1 | - | 1 | 220 |
