siRNA Details
Gene Symbol: 4930507D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027926.1 | 1 | 1825 | |||
GenBank mRNA | AK042417.1 | - | 1 | 1487 | ||
AK054359.1 | - | 1 | 1459 | |||
AK135283.1 | - | 1 | 1499 | |||
BC019469.1 | - | 1 | 1448 |
