siRNA Details
Gene Symbol: 4930507D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027926.1 | 1 | 1415 | |||
GenBank mRNA | AK015726.1 | - | 1 | 1416 | ||
AK042417.1 | - | 1 | 1077 | |||
AK054359.1 | - | 1 | 1049 | |||
AK135283.1 | - | 1 | 1089 | |||
BC019469.1 | - | 1 | 1038 |
