siRNA Details
Gene Symbol: Gm839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033190.1 | 2, 3 | 333 | |||
GenBank mRNA | AY512906.1 | - | 2, 3 | 233 | ||
BC147089.1 | - | 2, 3 | 333 | 9056153 | ||
BC147090.1 | - | 2, 3 | 333 | 9056152 |
