siRNA Details
Gene Symbol: Gm839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033190.1 | 1 | 42 | |||
GenBank mRNA | BC147089.1 | - | 1 | 42 | 9056153 | |
BC147090.1 | - | 1 | 42 | 9056152 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033190.1 | 1 | 42 | |||
GenBank mRNA | BC147089.1 | - | 1 | 42 | 9056153 | |
BC147090.1 | - | 1 | 42 | 9056152 |