siRNA Details
Gene Symbol: 2610001J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024619.1 | 3 | 653 | |||
GenBank mRNA | AK002763.1 | - | 3 | 636 | ||
AK011259.1 | - | 3 | 636 | |||
AK028169.1 | - | 3 | 654 | |||
AK035027.1 | - | 3 | 636 | |||
AK136158.1 | - | 3 | 637 | |||
AK152937.1 | - | 689 | ||||
BC070405.1 | - | 2 | 620 |

More Information
Other Species Targeted

Gene Ontology Categories:
