siRNA Details
Gene Symbol: C530005A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029450.1 | 2, 3 | 1898 | |||
GenBank mRNA | AK086654.1 | - | 2, 3 | 1900 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C530005A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029450.1 | 2, 3 | 1898 | |||
GenBank mRNA | AK086654.1 | - | 2, 3 | 1900 |