siRNA Details
Gene Symbol: C530005A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029450.1 | 1 | 471 | |||
GenBank mRNA | AK035052.1 | - | 1 | 476 | ||
AK039789.1 | - | 1 | 474 | |||
AK086654.1 | - | 1 | 473 | |||
AK140096.1 | - | 1 | 452 | |||
AK164083.1 | - | 1 | 436 |
