siRNA Details
Gene Symbol: 2810047C21Rik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015598.1 | 6 | 1148 | |||
GenBank mRNA | AK012911.1 | - | 653 | |||
AK028951.1 | - | 963 | ||||
AK045648.1 | - | 954 | ||||
AK083498.1 | - | 6 | 1149 | |||
BC071238.1 | - | 922 |
Gene Symbol: Gm3912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC026878.1 | - | 1 | 708 | 2615711 |
