siRNA Details
Gene Symbol: 2810047C21Rik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015598.1 | 2 | 430 | |||
GenBank mRNA | AK083498.1 | - | 2 | 431 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB654671.1 | - | 2 | 428 | Chr. 7 - 8088997 - 8093035 [-] on Build GRCm38 |
