siRNA Details
Gene Symbol: Gm14207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030683.1 | 4 | 646 | |||
GenBank mRNA | AK039958.1 | - | 4 | 647 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB201250.2 | - | 2 | 288 | Chr. 2 - 119321199 - 119324977 [-] on Build GRCm38 |
