siRNA Details
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015473.1 | 3 | 1533 | |||
NR_027832.1 | 3 | 1364 | ||||
GenBank mRNA | AK009929.1 | - | 3 | 1340 | ||
AK029148.1 | - | 1499 | ||||
AK030275.1 | - | 1496 | ||||
AK147270.1 | - | 1510 | ||||
BC026913.1 | - | 3 | 1330 |
