siRNA Details
Gene Symbol: D6Ertd474e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027803.1 | 4 | 2039 | |||
GenBank mRNA | AK139687.1 | - | 4 | 2035 | ||
BG081548.2 | - | 1 | 207 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D6Ertd474e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027803.1 | 4 | 2039 | |||
GenBank mRNA | AK139687.1 | - | 4 | 2035 | ||
BG081548.2 | - | 1 | 207 |