siRNA Details
Gene Symbol: 6330407A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028126.1 | 1 | 238 | |||
GenBank mRNA | AK018133.1 | - | 1 | 238 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6330407A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028126.1 | 1 | 238 | |||
GenBank mRNA | AK018133.1 | - | 1 | 238 |