siRNA Details
Gene Symbol: Scarna13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028576.1 | 1 | 162 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE633860.1 | - | 1 | 297 | Chr. 12 - 105031013 - 105031485 [-] on Build GRCm38 |
