siRNA Details
Gene Symbol: 2810454H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029441.1 | 1 | 702 | |||
GenBank mRNA | AK028630.1 | - | 1 | 702 | ||
AK051179.1 | - | 1 | 202 | |||
AK080385.1 | - | 1 | 269 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB639297.1 | - | 1 | 266 | Chr. 6 - 134898397 - 134899084 [+] on Build GRCm38 |
