siRNA Details
Gene Symbol: Gm12070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002890.1 | 1 | 113 | |||
GenBank mRNA | BC098095.1 | - | 1 | 113 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF732048.1 | - | 1 | 113 | Chr. 11 - 26785108 - 26786786 [+] on Build GRCm38 |
