siRNA Details
Gene Symbol: B930036N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154483.1 | - | 24 | |||
AK171461.1 | - | 1 | 24 |
Gene Symbol: Rnu5g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002852.3 | 1 | 23 | |||
GenBank mRNA | M10336.1 | - | 1 | 24 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: FQ226336, K00783.
Gene Ontology Categories:
