siRNA Details
Gene Symbol: Rnu5g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002852.3 | 1 | 44 | |||
GenBank mRNA | M10336.1 | - | 1 | 45 |
Gene Symbol: B930036N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047216.1 | - | 1 | 993 | ||
AK154483.1 | - | 45 | ||||
AK171461.1 | - | 1 | 45 |
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017789.1 | - | 45 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: FQ226336.
Gene Ontology Categories:
