siRNA Details
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033218.1 | 12 | 9468 | |||
NR_033615.1 | 10 | 6163 | ||||
GenBank mRNA | AK004146.1 | - | 1 | 1576 | ||
AK147260.1 | - | 12 | 9469 | |||
AK147653.1 | - | 12 | 9469 | |||
AY946044.1 | - | 10 | 7860 |
