siRNA Details
Gene Symbol: 1110038B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015536.1 | 4 | 390 | |||
NR_027943.1 | 5 | 537 | ||||
GenBank mRNA | AK002372.1 | - | 4 | 390 | ||
AK004150.1 | - | 5 | 538 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1110038B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015536.1 | 4 | 390 | |||
NR_027943.1 | 5 | 537 | ||||
GenBank mRNA | AK002372.1 | - | 4 | 390 | ||
AK004150.1 | - | 5 | 538 |