siRNA Details
Gene Symbol: Gm7337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003652.2 | 1 | 1003 | |||
GenBank mRNA | AK133184.1 | - | 1 | 1004 | ||
BC147596.1 | - | 1 | 927 | 9056586 | ||
BC147623.1 | - | 1 | 927 | 9056613 |
