siRNA Details
Gene Symbol: Gm13749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027824.1 | 2 | 392 | |||
GenBank mRNA | AK081719.2 | - | 2 | 394 | ||
BC147571.1 | - | 2 | 363 | 9056561 | ||
BC147573.1 | - | 2 | 363 | 9056563 |
