siRNA Details
Gene Symbol: 1700066O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015541.2 | 2 | 662 | |||
GenBank mRNA | AK015660.1 | - | 2 | 663 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700066O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015541.2 | 2 | 662 | |||
GenBank mRNA | AK015660.1 | - | 2 | 663 |